Sartorius YTM10MA Temperature Calibration Bowl for Mark-3 LTE Moisture Analyzer
Representative image only.
Sartorius YTM10MA Temperature Calibration Bowl for Mark-3 LTE Moisture Analyzer
Sartorius | Mfr: YTM10MA| Item: 1121327
Provides automatic calibartions
₹211,000.00INR / Each
₹211,000.00INR / Each
Product Description
Key Features
  • Designed for use with Mark 3 Series moisture analyzers
  • 1 year warranty
More About this Item
The temperature calibration bowl provides automatic calibrations for your moisture analyzer. To calibrate the internal temperature sensor, simply remove the tray support and breeze shield wihin the heating chamber, and install the bowl.
1 Year Warranty
Specifications
  • DescriptionTemperature Calibration Bowl for Mark-3 LTE Moisture Analyzer