Sartorius Secura® NTEP-Approved Toploading Balances
Designed for fast, highly accurate weighing results
  • NTEP approved legal for trade
  • Password security offers protection against unauthorized access
  • Optoelectronic level sensor provides continuous real-time monitoring
  • Automatic internal calibration and adjustment
  • Touch-screen display with graphical user interface
Providing highly accurate weighing results and operating convenience, the Secura® toploading balances feature built-in protection systems for complete reliability and regulatory compliance. The isoCAL adjustment function alerts you whenever a defined interval has elapsed, or a temperature limit has been exceeded. The balance then performs fully automatic span adjustment using its internal weight. An internal Cal Audit Trail function stores each calibration and adjustment procedure, generating a 100% traceable audit trail for your quality assurance. A mini USB interface and Plug & Play technology enables communication with a PC or printer.
A continuous real-time level monitoring function prevents you from reporting inaccurate results due to an unleveled balance. It alerts you immediately, and temporarily blocks data transfer from going to a printer or computer to ensure data integrity.
The balances also feature a SQmin function that checks your sample quantity to confirm it is within the acceptable USP operating range. Weighing applications include general weighing, density, percentage, check weighing, peak hold, counting, unstable conditions, and animal weighing.
780,000.00 - ₹1,731,000.00INR / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Calibration Type Capacity (g) Readability (mg)
Availability Pricing
Sartorius SECURA1103-1NUS NTEP-Certified Secura Toploading Balance 1100g x 0.001g Internal Calibration 1180030 Mfr # SECURA1103-1NUS
COMPARE
Internal 1100 1
₹1,731,000.00INR / Each
Sartorius SECURA613-1NUS NTEP- Certified Secura Toploading Balance 610g  x 0.001g Internal Calibration 1180031 Mfr # SECURA613-1NUS
COMPARE
Internal 610 1
₹1,021,000.00INR / Each
Sartorius SECURA313-1NUS NTEP-Certified Secura Toploading Balance 310g  x 0.001g Internal Calibration 1180032 Mfr # SECURA313-1NUS
COMPARE
Internal 310 1
₹893,000.00INR / Each
Sartorius SECURA6102-1NUS NTEP-Certified Secura Toploading Balance 6100g x 0.01g, Internal Calibration 1180033 Mfr # SECURA6102-1NUS
COMPARE
Internal 6100 10
₹1,050,000.00INR / Each
Sartorius SECURA3102-1NUS NTEP-Certified Secura Toploading Balance 3100g x 0.01g Internal Calibration 1180034 Mfr # SECURA3102-1NUS
COMPARE
Internal 3100 10
₹1,005,000.00INR / Each
Sartorius SECURA1102-1NUS NTEP-Certified Secura Toploading Balance 1100g x 0.01g Internal Calibration 1180035 Mfr # SECURA1102-1NUS
COMPARE
Internal 1100 10
₹780,000.00INR / Each
Sartorius SECURA6101-1NUS NTEP-Certified Secura Toploading Balance 6100g x 0.1g Internal Calibration 1180036 Mfr # SECURA6101-1NUS
COMPARE
Internal 6100 100
₹859,000.00INR / Each
Showing 1- 7 of 7