Sartorius® Quintix Pro Analytical Balances
Designed for precision and usability - suitable for wide range of laboratory applications
  • Motorized feet simplify leveling
  • Easy access to menu option with 7” high resolution touch display
  • Automatic internal adjustment (isoCAL) delivers accurate results
  • Stand-by or switch-off modes saves energy
  • Chemically-resistant materials offer easy cleaning and maintenance
  • Automatic dim function reduces display brightness, saves power
  • Built-in interactive manual allows quick access to information
The Quintix Pro analytical balances are designed for usability and precision, making them ideal for a variety of laboratory applications. They feature a 7-inch high-resolution touch display with a user-friendly interface, ensuring easy navigation. Key functionalities include an internal motorized leveling system and automatic internal adjustment (isoCAL) for optimal accuracy. The LevelControl function monitors balance stability in real-time, while the semi-automatic filter adaptation (AFA) allows customization for specific tasks.
Data transfer is seamless, with multiple connection options including three USB Type C interfaces, RS-232, and ethernet. The balances support GLP/GMP-compliant printouts and facilitate data transfer to USB or databases.
These balances are versatile enough for cleanrooms (grade A and B) and specialized environments like vacuum, argon, and nitrogen settings. They come with an extended language package (15 languages) and 15 integrated application programs for various weighing tasks. Security features include customizable access levels with passcode protection.
Additional features, such as stand-by mode, automatic dimming, and a user-defined start timer, promote energy efficiency. Made with recycled materials and eco-friendly packaging, the Quintix Pro balances prioritize sustainability without compromising on performance, making them an excellent choice for any laboratory setting.
1,127,000.00 - ₹1,678,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Readability (mg) Description
Availability Pricing
Sartorius Quintix Pro QTX324IRU-1S Analytical Balance, 320g x 0.1mg, Internal Calibration and Guided Leveling 1197699 Mfr # QTX324IRU-1S
COMPARE
320 0.1 Analytical Balance, 320g x 0.1mg, with Internal Calibration and Guided Leveling
₹1,486,000.00INR / Each
Sartorius Quintix Pro QTX224IRU-1S Analytical Balance, 220g x 0.1mg, Internal Calibration and Guided Leveling 1197700 Mfr # QTX224IRU-1S
COMPARE
220 0.1 Analytical Balance, 220g x 0.1mg with Internal Calibration and Guided Leveling
₹1,294,000.00INR / Each
Sartorius Quintix Pro QTX124IRU-1S Analytical Balance, 120g x 0.1mg, Internal Calibration and Guided Leveling 1197701 Mfr # QTX124IRU-1S
COMPARE
120 0.1 Analytical Balance, 120g x 0.1mg, with Internal Calibration and Guided Leveling
₹1,127,000.00INR / Each
Sartorius Quintix Pro QTX324IMU-1S Analytical Balance, 320g x 0.1mg, Internal Calibration and Motorized Leveling 1197717 Mfr # QTX324IMU-1S
COMPARE
320 0.1 Analytical Balance, 320g x 0.1mg, with Internal Calibration and Motorized Leveling
₹1,678,000.00INR / Each
Sartorius Quintix Pro QTX224IMU-1S Analytical Balance, 220g x 0.1mg, Internal Calibration and Motorized Leveling 1197718 Mfr # QTX224IMU-1S
COMPARE
220 0.1 Analytical Balance, 220g x 0.1mg, with Internal Calibration and Motorized Leveling
₹1,486,000.00INR / Each
Sartorius Quintix Pro QTX124IMU-1S Analytical Balance, 120g x 0.1mg, Internal Calibration and Motorized Leveling 1197719 Mfr # QTX124IMU-1S
COMPARE
120 0.1 Analytical Balance, 120g x 0.1mg, with Internal Calibration and Motorized Leveling
₹1,318,000.00INR / Each