Sartorius Cubis II (MCE) Analytical Balances
Individual configuration of draft shield and integrated ionizer provides adaptation to any weighing task
  • Monolithical weighing system, integrated climate sensors, individual sample holders offer industry leading performance Individual QApp workflows and motorized auto-leveling reduce error 21 CFR Part 11 compliance, integrated audit-trail, state-of-the-art user management provides excellent data integrity Innovative TFT touch screen operation Factory installed set of essential weighing applications Essential user interface features high-contrast touch display for easy operation
Analytical balances have a maximum capacity between 120 g and 520 g. The automatic glass draft shield with integrated ionizer removes interfering electrostatic charges on samples and sample containers. The manual glass draft shield has smooth-action doors. Large, high-contrast touch display with factory-installed essential weighing applications for easy operation.
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (g) Description
Availability Pricing
Sartorius Cubis II Analytical Balance, 520 g x 0.1 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz 3974230 Mfr # MCE524S-2S00-I
COMPARE
520 Analytical Balance, 520 g x 0.1 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz
₹4,708,000.00INR / Each
Sartorius Cubis II Analytical Balance, 320 g x  0.1 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz 3974232 Mfr # MCE324S-2S00-I
COMPARE
320 Analytical Balance, 320 g x 0.1 mg, with Automatic Glass Draft Shield and Integrated Ionizer; 100 to 240 VAC, 50/60 Hz
₹4,264,000.00INR / Each