Sartorius Arium® Pro Ultrapure Water Systems
Each modular designed system focuses on the most important functions to produce high-quality ultrapure water
  • Each system meets and exceeds the ASTM Type I standard
  • The lowest TOC level, along with a real-time monitoring function
  • Exceptionally retention rates for endotoxins, DNase, and RNase
  • Glass display with touch-activated functions and intuitive menu navigation
  • “Favorites” function for direct access to preprogrammed volume dispensing
The Arium Pro system provides the ultimate flexibility at an excellent cost-benefit ratio while providing the highest quality ultrapure water. Dispense up to 2 L/min of ASTM Type I ultrapure water with conductivity of 0.055 µS/cm (18.2 MO x cm), bacterial content of < 0.001 CFU/mL*, and no particles > 0.22 µm*. Select from five Arium Pro series water purification systems, with models featuring an ultrafilter, UV lamp, or TOC monitor to meet your application needs.
All models feature "Favorite" function for quick access to preprogrammed dispensing volume that saves time and minimizes errors. Graphic and acoustic signals alert you when limits are exceeded or data falls below minimum limits. Easily save your data by connecting to a printer using the RS-232 port.
Call For Price
14 variations of this product are available.
See All Product Options Below
14 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Special Features With Ultrafilter With UV Lamp
Availability Pricing
Sartorius Arium® Pro Basic Wall-Mount System 9929340 Mfr # H2OBASIC-B
COMPARE
Wall mount No No
Call For Price
Sartorius Arium® Pro Basic Benchtop System 9929341 Mfr # H2OBASIC-T
COMPARE
Benchtop No No
Call For Price
Sartorius Arium® Pro DI Wall-Mount System 9929372 Mfr # H2OPRO-DI-B
COMPARE
Wall mount No No
Call For Price
Sartorius Arium® Pro DI Benchtop System 9929373 Mfr # H2OPRO-DI-T
COMPARE
Benchtop No No
Call For Price
Sartorius Arium® Pro UF Wall-Mount System 9929374 Mfr # H2OPRO-UF-B
COMPARE
Wall mount and ultrafilter Yes No
Call For Price
Sartorius Arium® Pro UF Benchtop System 9929375 Mfr # H2OPRO-UF-T
COMPARE
Benchtop and ultrafilter Yes No
Call For Price
Sartorius Arium® Pro UV Wall-Mount System 9929376 Mfr # H2OPRO-UV-B
COMPARE
Wall mount and UV lamp No Yes
Call For Price
Sartorius Arium® Pro UV Wall-Mount System with TOC Monitor 9929377 Mfr # H2OPRO-UV-B-TOC
COMPARE
Wall mount, UV lamp, and TOC monitor No Yes
Call For Price
Sartorius Arium® Pro UV Benchtop System 9929378 Mfr # H2OPRO-UV-T
COMPARE
Benchtop and UV lamp No Yes
Call For Price
Sartorius Arium® Pro UV Benchtop System with TOC Monitor 9929379 Mfr # H2OPRO-UV-T-TOC
COMPARE
Benchtop, UV lamp, and TOC monitor No Yes
Call For Price
Sartorius Arium® Pro VF Wall-Mount System 9929380 Mfr # H2OPRO-VF-B
COMPARE
Wall mount, ultrafilter, and UV lamp Yes No
Call For Price
Sartorius Arium® Pro VF Wall-Mount System with TOC Monitor 9929381 Mfr # H2OPRO-VF-B-TOC
COMPARE
Wall mount, ultrafilter, UV lamp, and TOC monitor Yes No
Call For Price
Sartorius Arium® Pro VF Benchtop System 9929382 Mfr # H2OPRO-VF-T
COMPARE
Benchtop, ultrafilter, and UV lamp Yes No
Call For Price
Sartorius Arium® Pro VF Benchtop System with TOC Monitor 9929383 Mfr # H2OPRO-VF-T-TOC
COMPARE
Benchtop, ultrafilter, UV lamp, and TOC monitor Yes No
Call For Price
Showing 1- 14 of 14