Sani-Tech® Tee Fittings, PVDF
Effortlessly integrate tee fittings into high-purity systems
  • Seamlessly connect and branch flow paths with precision-engineered tee fittings
  • Crafted from high-purity PVDF (polyvinylidene fluoride) for superior chemical and thermal resistance
  • Exceptional resistance to corrosion, pitting, and UV radiation ensures long-term reliability
  • Meets 21 CFR 177.2510, USP Class VI, 3-A, NSF-61, and USDA standards for ultimate safety and compliance
Sani-Tech® tee fittings provide an optimal solution for industries demanding the highest levels of purity. With outstanding surface smoothness, low extractables, and minimal TOC levels, these fittings help maintain system integrity while preventing contamination. Designed to withstand aggressive chemicals and operating temperatures up to 175°F, they outperform traditional materials in pharmaceutical, semiconductor, DI water, and food & beverage applications.
16,400.00 - ₹20,700.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
Sani-Tech®  Sani-Pro® K Sanitary Fitting, Tee, PVDF, 3/4" Mini Tri-Clamp; 1/Each 5013501 Mfr # KT075
COMPARE
PVDF 3/4
₹16,400.00INR / Each
Sani-Tech®  Sani-Pro® K Sanitary Fitting, Tee, PVDF, 1-1/2" Tri-Clamp; 1/Each 5013504 Mfr # KT150
COMPARE
PVDF 1-1/2
₹20,700.00INR / Each