Sani-Tech® Sani-Pro® K Sanitary Fitting, Tee, PVDF, 3/4" Mini Tri-Clamp; 1/Each
Representative image only.
Sani-Tech® Sani-Pro® K Sanitary Fitting, Tee, PVDF, 3/4" Mini Tri-Clamp; 1/Each
SANI-TECH | Mfr: KT075| Item: 5013501
Effortlessly integrate tee fittings into high-purity systems
₹16,400.00INR / Each
₹16,400.00INR / Each
Buy It Together
Current Item
Sani-Tech®  Sani-Pro® K Sanitary Fitting, Tee, PVDF, 3/4" Mini Tri-Clamp; 1/Each
Sani-Tech® Sani-Pro® K Sanitary Fitting, Tee, PVDF, 3/4" Mini Tri-Clamp; 1/Each
5013501
₹16,400.00INR / Each
+ + =
Total Price 27800.00 ADD 3 ITEMS TO CART
Customers Also Bought
Product Description
Includes
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Key Features
  • Seamlessly connect and branch flow paths with precision-engineered tee fittings
  • Crafted from high-purity PVDF (polyvinylidene fluoride) for superior chemical and thermal resistance
  • Exceptional resistance to corrosion, pitting, and UV radiation ensures long-term reliability
  • Meets 21 CFR 177.2510, USP Class VI, 3-A, NSF-61, and USDA standards for ultimate safety and compliance
More About this Item
Sani-Tech® tee fittings provide an optimal solution for industries demanding the highest levels of purity. With outstanding surface smoothness, low extractables, and minimal TOC levels, these fittings help maintain system integrity while preventing contamination. Designed to withstand aggressive chemicals and operating temperatures up to 175°F, they outperform traditional materials in pharmaceutical, semiconductor, DI water, and food & beverage applications.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Connection TypeSanitary
  • Sanitary Clamp Size (Port 1) (InchShort)3/4
  • Sanitary Clamp Size (Port 2) (in)3/4
  • Sanitary Clamp Size (Port 3) (InchShort)3/4
  • Flow PatternTee
  • Max Temperature (° F)175
  • Max Temperature (° C)79.4
  • DescriptionSanitary Fitting, Tee, PVDF, 3/4" Mini Tri-Clamp; 1/Each