Sani-Tech® Sanitary Clamp 90° Elbows, Polypropylene
Seamless, corrosion-free flow for critical applications
  • Optimized fluid flow with sweep elbow design that minimizes pressure drop and flow restrictions
  • Beadless fusion technology SIB® (Smooth Inner Bore) eliminates entrapment areas, reducing biofilm risks
  • Seamless integration with stainless steel – fitting dimensions match ASME BPE standards
  • Chemical and corrosion resistance with high-purity polypropylene(PP)
  • Easy installation with simple fabrication and laser-etched lot traceability
Sani-Tech® High Purity Piping Elbows are ideal for pharmaceutical, biotech, and food & beverage applications, including WFI distribution, dialysis water systems, and metal-sensitive product transfers. With superior chemical resistance and smooth inner surfaces, these elbows eliminate contamination risks and minimize maintenance. The advanced SIB® beadless fusion ensures a seamless, obstruction-free flow path, making installation and inspection effortless.
9,300.00 - ₹27,800.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (InchShort)
Availability Pricing
Sani-Tech® T Sanitary Fitting, 90° Elbow, Polypropylene, 3/4" Mini Tri-Clamp; 1/Each 5013497 Mfr # T90075
COMPARE
Polypropylene 3/4
₹11,000.00INR / Each
Sani-Tech® T Sanitary Fitting, 90° Elbow, Polypropylene, 1" Ladish Tri-Clamp; 1/Each 5013498 Mfr # T90100L
COMPARE
Polypropylene 1
₹27,800.00INR / Each
Sani-Tech® T Sanitary Fitting, 90° Elbow, Polypropylene, 1-1/2" Tri-Clamp; 1/Each 5013499 Mfr # T90150
COMPARE
Polypropylene 1-1/2
₹9,300.00INR / Each