Sani-Tech® T Sanitary Fitting, 90° Elbow, Polypropylene, 1" Ladish Tri-Clamp; 1/Each
Representative image only.
Sani-Tech® T Sanitary Fitting, 90° Elbow, Polypropylene, 1" Ladish Tri-Clamp; 1/Each
SANI-TECH | Mfr: T90100L| Item: 5013498
Seamless, corrosion-free flow for critical applications
₹27,800.00INR / Each
₹27,800.00INR / Each
Product Description
Key Features
  • Optimized fluid flow with sweep elbow design that minimizes pressure drop and flow restrictions
  • Beadless fusion technology SIB® (Smooth Inner Bore) eliminates entrapment areas, reducing biofilm risks
  • Seamless integration with stainless steel – fitting dimensions match ASME BPE standards
  • Chemical and corrosion resistance with high-purity polypropylene(PP)
  • Easy installation with simple fabrication and laser-etched lot traceability
More About this Item
Sani-Tech® High Purity Piping Elbows are ideal for pharmaceutical, biotech, and food & beverage applications, including WFI distribution, dialysis water systems, and metal-sensitive product transfers. With superior chemical resistance and smooth inner surfaces, these elbows eliminate contamination risks and minimize maintenance. The advanced SIB® beadless fusion ensures a seamless, obstruction-free flow path, making installation and inspection effortless.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPolypropylene
  • Sanitary Clamp Size (Port 1) (InchShort)1
  • Sanitary Clamp Size (Port 2) (in)1
  • Max Temperature (° F)125
  • DescriptionSanitary Fitting, 90° Sweep Elbow, Polypropylene, 1" Ladish Tri-Clamp; 1/Each