Sani-Tech® Pipe Thread Adapters
Provide a seamless connection between sanitary ferrules and standard pipe threads, ensuring secure and contamination-free fluid transfer
  • Easily connect sanitary ferrules to male or female pipe threads
  • Choose polypropylene (PP) for moderate service applications or PVDF for superior chemical and high-temperature resistance
  • Both materials resist corrosion, pitting, and scaling, ensuring long-lasting performance
Manufactured from high-purity thermoplastics, these adapters are available in two materials to meet a variety of application needs. Polypropylene (PP) offers excellent chemical resistance, durability, and non-conductivity, making it ideal for pharmaceutical, food & beverage, dairy, and RO/DI water systems with an operating temperature of up to 125°F. For more demanding environments, PVDF provides superior chemical resistance, UV stability, and low extractables, making it the preferred choice for biotechnology, semiconductor, USP water systems, and WFI water systems, withstanding temperatures up to 175°F.
36,900.00 - ₹48,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Fitting Material Sanitary Clamp Size (Port 1) (InchShort) Thread Size NPT(M) (Port 1) (InchShort)
Availability Pricing
Sani-Tech® Sanitary to Threaded Adapter, Straight Adapter, Polypropylene, 1" Ladish Tri-Clamp x 1/2" NPT(M); 1/Each 5013482 Mfr # TAD100L-08M
COMPARE
Polypropylene 1 1/2
₹36,900.00INR / Each
Sani-Tech® Sanitary to Threaded Adapter, Straight Adapter, PVDF, 1" Ladish Tri-Clamp x 1/2" NPT(M); 1/Each 5013485 Mfr # KAD100L-08M
COMPARE
PVDF 1 1/2
₹48,000.00INR / Each
Sani-Tech® Sanitary to Threaded Adapter, Straight Adapter, PVDF, 1-1/2" Tri-Clamp x 1-1/2" NPT(M); 1/Each 5013487 Mfr # KAD150-24M
COMPARE
PVDF 1-1/2 1 1/2
₹38,500.00INR / Each