Sani-Tech® Sanitary to Threaded Adapter, Straight Adapter, PVDF, 1" Ladish Tri-Clamp x 1/2" NPT(M); 1/Each
Representative image only.
Sani-Tech® Sanitary to Threaded Adapter, Straight Adapter, PVDF, 1" Ladish Tri-Clamp x 1/2" NPT(M); 1/Each
SANI-TECH | Mfr: KAD100L-08M| Item: 5013485
Provide a seamless connection between sanitary ferrules and standard pipe threads, ensuring secure and contamination-free fluid transfer
₹48,000.00INR / Each
₹48,000.00INR / Each
Product Description
Key Features
  • Easily connect sanitary ferrules to male or female pipe threads
  • Choose polypropylene (PP) for moderate service applications or PVDF for superior chemical and high-temperature resistance
  • Both materials resist corrosion, pitting, and scaling, ensuring long-lasting performance
More About this Item
Manufactured from high-purity thermoplastics, these adapters are available in two materials to meet a variety of application needs. Polypropylene (PP) offers excellent chemical resistance, durability, and non-conductivity, making it ideal for pharmaceutical, food & beverage, dairy, and RO/DI water systems with an operating temperature of up to 125°F. For more demanding environments, PVDF provides superior chemical resistance, UV stability, and low extractables, making it the preferred choice for biotechnology, semiconductor, USP water systems, and WFI water systems, withstanding temperatures up to 175°F.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF
  • Connection TypeSanitary clamp to threaded fitting
  • Thread Size NPT(M) (Port 1) (InchShort)1/2
  • Sanitary Clamp Size (Port 1) (InchShort)1
  • Flow PatternStraight through
  • DescriptionSanitary to Threaded Adapter, Straight Adapter, PVDF, 1" Ladish Tri-Clamp x 1/2" NPT(M); 1/Each