Saint-Gobain Pure-Fit® Tubing Clamps, PP and PVDF
No sharp edges or corners, minimizing potential for snags and punctures
  • Install on or over a completed assembly
  • Provides complete flow stoppage
  • Locking mechanism ensures against unwanted release
  • Easy single-handed installation and operation
  • USP <88> Class VI compliant
Uniquely designed Pure-Fit tubing clamps allow for easy installation over tubing in a completed assembly. Clamps feature smooth edges, eliminating any chances of snags, scratches, and punctures. Use the side release mechanism for single-handed installation and operation. A fully encapsulated press-down locking mechanism minimizes any chances of accidental release. For total lock out, thread a cable tie through bore hole.
21,400.00 - ₹69,500.00INR / Pkg of 25
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Clamp Material Tubing Size Range
Availability Pricing
Saint Gobain Pure-Fit® Tubing Clamps, PP, 1/8" to 3/8"; 25/Pk 5014353 Mfr # PFTC375
COMPARE
Polypropylene 1/8" to 3/8" OD
₹21,400.00INR / Pkg of 25
Saint Gobain Pure-Fit® Tubing Clamps, PP, 3/8" to 3/4"; 25/Pk 5014354 Mfr # PFTC750
COMPARE
Polypropylene 3/8" to 3/4" OD
₹26,100.00INR / Pkg of 25
Saint Gobain Pure-Fit® Tubing Clamps, PVDF, 3/8" to 3/4"; 25/Pk 5014355 Mfr # PFTC750PVDF
COMPARE
PVDF 3/8" to 3/4" OD
₹69,500.00INR / Pkg of 25
Saint Gobain Pure-Fit® Tubing Clamps, PVDF, 1/8" to 3/8"; 25/Pk 5014356 Mfr # PFTC375PVDF
COMPARE
PVDF 1/8" to 3/8" OD
₹39,300.00INR / Pkg of 25