Saint Gobain Pure-Fit® Tubing Clamps, PVDF, 1/8" to 3/8"; 25/Pk
Representative image only.
Saint Gobain Pure-Fit® Tubing Clamps, PVDF, 1/8" to 3/8"; 25/Pk
Saint Gobain | Mfr: PFTC375PVDF| Item: 5014356
No sharp edges or corners, minimizing potential for snags and punctures
₹39,300.00INR / Pkg of 25
₹39,300.00INR / Pkg of 25
Product Description
Key Features
  • Install on or over a completed assembly
  • Provides complete flow stoppage
  • Locking mechanism ensures against unwanted release
  • Easy single-handed installation and operation
  • USP <88> Class VI compliant
  • PVDF construction
  • Sterilize by autoclave or gamma irradiation
  • Maximum recommended working temperature is 130°C (266°F)
  • Accommodates 1/8" to 3/8" OD tubing
  • Manufacturer defect warranty
More About this Item
Uniquely designed Pure-Fit tubing clamps allow for easy installation over tubing in a completed assembly. Clamps feature smooth edges, eliminating any chances of snags, scratches, and punctures. Use the side release mechanism for single-handed installation and operation. A fully encapsulated press-down locking mechanism minimizes any chances of accidental release. For total lock out, thread a cable tie through bore hole.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Clamp MaterialPVDF
  • Tubing Size Range1/8" to 3/8" OD
  • DescriptionSaint-Gobain Pure-Fit® Tubing Clamps, PVDF, 1/8" to 3/8"; 25/Pk
  • WarrantyManufacturer defect warranty