Rotary Vane Vacuum Pumps
Highest reliability for stable operation and long product lifecycle
  • Reduced risk of chemical attacks and oil breakdown by diluting chemical vapours with a larger oil chamber
  • Ensures protection from sublimed chemical vapours with coating on both pump and oil case
  • Enhanced air flow allows the pump to run 10°C cooler than standard rotary vane pumps
Robust pumps have a low noise level, making them exceptionally quiet for undisturbed work. Forced oil lubrication ensures that moving parts are bathed in fresh oil, minimizing breakdown. Anti-suck back valve prevents oil from being sucked into application in the event of a power failure.
379,000.00 - ₹527,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Free-Air Capacity (CFM) Oil Capacity (Liters) Description
Availability Pricing
Welch CRV Pro 8 Rotary Vane Pump, 5.6 cfm, 0.002 mbar, 0.370 kw, with flow rate 63 L/min 1634966 Mfr # 3082-00
COMPARE
5.6 1 Welch CRV Pro 8 Rotary Vane Pump, 5.6 cfm, 0.002 mbar, 0.370 kw, with flow rate 63 L/min
₹527,000.00INR / Each
Welch CRV Pro 4 Rotary Vane Pump, 2.8 cfm, 0.002 mbar, with flow rate 122 L/min 1634973 Mfr # 3042-00
COMPARE
2.8 1.15 Welch CRV Pro 4 Rotary Vane Pump, 2.8 cfm, 0.002 mbar, with flow rate 122 L/min
₹379,000.00INR / Each
Welch CRV Pro 6 Rotary Vane Pump, 3.7 cfm, 0.002 mbar, with flow rate 85 L/min 1634975 Mfr # 3062-00
COMPARE
3.7 1.15 Welch CRV Pro 6 Rotary Vane Pump, 3.7 cfm, 0.002 mbar, with flow rate 85 L/min
₹437,000.00INR / Each