ResinTech HYDRA-DI Deionized Water Systems
Turn-key system provides on-demand service at an economical price
  • Can be wall mounted and offers low profile
  • Low resistivity indicating light for cartridge replacement
  • Economically produces DI water
  • Turn-key system for on demand service
  • Designed for city water and tap water supply
  • Includes outlet isolation valve, spanner wrench and associated tubing
  • Made in the USA!
The Hydra DI System is a pre-assembled cartridge system that provides deionized water using a staged filtration process. Water purification is provided using a three-step process. Stage 1 removes particles greater than 5 micron, chlorine, and organics. Stages 2 and 3 are deionizers designed to remove dissolved organics and silica to trace levels.
As the water exits the system, a resistivity indicator provides feedback on system performance. Green light indicates water purity of the set point. A red light indicates it is time to change the cartridges. These premium grade resins are specially designed and processed to shed low levels of organics. The Hydra DI series includes all cartridges and resistivity light for a total turnkey system. The system configuration uses mixed bed cartridges for applications requiring TYPE II water quality.
These low profile, wall-mounted water systems are available in high purity, high capacity, and low TOC configurations and feature a pre-assembled cartridge arrangement with either 2½" or 4½" diameter filter housing canisters.
256,000.00 - ₹457,000.00INR / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Flow Rate (GPH) Capacity Resistivity Threshold
Availability Pricing
ResinTech HYS-122-S-HP Hydra "Slimline" High Purity DI System 9928511 Mfr # HYS-122-S-HP
COMPARE
30 1800 grains as CaCO3 200 KΩ
₹256,000.00INR / Each
ResinTech HYS-122-B-HP Hydra "Big Blue" High Purity DI System 9928512 Mfr # HYS-122-B-HP
COMPARE
78 4760 grains as CaCO3 200 KΩ
₹457,000.00INR / Each
ResinTech HYS-122-S-HC Hydra "Slimline" High Capacity DI System 9928513 Mfr # HYS-122-S-HC
COMPARE
30 2160 grains as CaCO3 20 KΩ
₹256,000.00INR / Each
ResinTech HYS-122-B-HC Hydra "Big Blue" High Capacity DI System 9928514 Mfr # HYS-122-B-HC
COMPARE
78 5710 grains as CaCO3 20 KΩ
₹457,000.00INR / Each