Replacement Electrodes
Low maintenance gel-filled electrodes suitable for measuring free chlorine, chlorine dioxide and ozone
  • Electrode sensing material--gold
  • Ag/AgCl/T epoxy gel reference
  • Max pressure-16 bar @ 20°
  • Max temperature 70° C
Electrode 27400-05 is a replacement for std systems (for 27400-01 and -07).
Electrode 27400-06 is a replacement for the self cleaning system (for 27400-03 and -09)
119,000.00 - ₹189,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Kuntze 24135130K Free Chlorine Disinfectant Analyzer Replacement Electrode 2740005 Mfr # 24135130K
COMPARE
Free Chlorine Disinfectant Analyzer Replacement Electrode
₹119,000.00INR / Each
Kuntze 24135140K Free Chlorine Replacement Electrode for Self Cleaning 2740006 Mfr # 24135140K
COMPARE
Free Chlorine Replacement Electrode for Self Cleaning
₹189,000.00INR / Each