Kuntze 24135130K Free Chlorine Disinfectant Analyzer Replacement Electrode
Representative image only.
Kuntze 24135130K Free Chlorine Disinfectant Analyzer Replacement Electrode
Kuntze | Mfr: 24135130K| Item: 2740005
Low maintenance gel-filled electrodes suitable for measuring free chlorine, chlorine dioxide and ozone
₹119,000.00INR / Each
₹119,000.00INR / Each
Product Description
Key Features
  • Electrode sensing material--gold
  • Ag/AgCl/T epoxy gel reference
  • Max pressure-16 bar @ 20°
  • Max temperature 70° C
More About this Item
Electrode 27400-05 is a replacement for std systems (for 27400-01 and -07).
Electrode 27400-06 is a replacement for the self cleaning system (for 27400-03 and -09)
Conformite Europeenne
Specifications
  • Control TypeOn/Off
  • Min Dissolved Oxygen (mg/L (ppm))0
  • Max Dissolved Oxygen (mg/L (ppm))20
  • DescriptionFree Chlorine Disinfectant Analyzer Replacement Electrode