Raytek® GP-Series Infrared Temperature Sensors
Get noncontact measurement of your system
  • Laser-sighted sensors are perfect for taking measurements in hard to reach locations
  • For small or distant targets
Choose from sensors with standard distance-to-target size ratios or with a non-laser close focus for small close-range measurement spots. Laser shuts off automatically ambient temperatures greater than 120°F (50°C). Order sensors with an optional air-cooling jacket to operate in environments with ambient temperatures up to 350°F (177°C).
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Distance To Target Size Ratio (D:S) Max Temperature (° F) Description
Availability Pricing
Raytek Standard Focus, Non-Laser, Infrared Temperature Sensor 3967102 Mfr # RAYGPRSF
COMPARE
50:1 150 Standard Focus, Non-Laser, Infrared Temperature Sensor
₹755,000.00INR / Each
Raytek Close Focus, Non-Laser, Infrared Temperature Sensor 3967104 Mfr # RAYGPRCF
COMPARE
35:1 150 Close Focus, Non-Laser, Infrared Temperature Sensor
₹755,000.00INR / Each
Raytek Standard Focus, Laser-Sighted, Infrared Temperature Sensor 3967120 Mfr # RAYGPSSFL
COMPARE
50:1 150 Standard Focus, Laser-Sighted, Infrared Temperature Sensor