QLA Zymark-Compatible Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is acid etched on each vessel Glass vessels offer good thermal conductivity Highly resistance to scratching
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom.
62,300.00 - ₹87,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Color Material
Availability Pricing
QLA Standard Vessel with Centering Ring for Zymark/MultiDose®, 1000 mL, Amber Glass; Serialized 4130644 Mfr # GLA9CR-AZM
COMPARE
1000 Amber Glass
₹87,200.00INR / Each
QLA Standard Vessel with Centering Ring for Zymark/MultiDose®, 1000 mL, Clear Glass; Serialized 4130646 Mfr # GLA9CR-ZM
COMPARE
1000 Clear Glass
₹62,300.00INR / Each