QLA Standard Vessel with Centering Ring for Zymark/MultiDose®, 1000 mL, Amber Glass; Serialized
Representative image only.
QLA Standard Vessel with Centering Ring for Zymark/MultiDose®, 1000 mL, Amber Glass; Serialized
| Mfr: GLA9CR-AZM| Item: 4130644
QLA vessels provide more precise and consistent dissolution results
₹87,200.00INR / Each
₹87,200.00INR / Each
Product Description
Key Features
  • Unique serial number is acid etched on each vessel Glass vessels offer good thermal conductivity Highly resistance to scratching
  • Amber glass is ideal for light-sensitive compounds
More About this Item
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom.
.
Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • ColorAmber
  • DescriptionStandard Vessel with Centering Ring for Zymark/MultiDose®, 1000 mL, Amber Glass; Serialized