QLA Vessels Compatible with Agilent, VanKel, and Varian Dissolution Apparatus
QLA vessels provide a more precise and consistent dissolution experience
  • Compatible with Agilent, VanKel, and Varian dissolution apparatus
Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.
47,000.00 - ₹154,000.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottle Color Capacity (mL) Special Features
Availability Pricing
QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 2000 mL, Clear Glass; Serialized 4130600 Mfr # GLA02K-01
COMPARE
Clear 2000 Standard
₹88,500.00INR / Each
QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 500 mL; Clear Glass 4130614 Mfr # GLA500-01
COMPARE
Clear 500 Standard
₹53,800.00INR / Each
QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 500 mL; Amber Glass 4130616 Mfr # GLA500-AV
COMPARE
Amber 500 Standard
₹109,000.00INR / Each
QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Clear Glass; Serialized 4130619 Mfr # GLA900-01
COMPARE
Clear 1000 Standard
₹47,000.00INR / Each
QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Amber Glass; Serialized 4130629 Mfr # GLA900-AV
COMPARE
Amber 1000 Standard
₹88,700.00INR / Each
QLA PEAK Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Clear Glass; Serialized 4130647 Mfr # GLA9PK-01
COMPARE
Clear 1000 PEAK style
₹68,700.00INR / Each
QLA PEAK Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Amber Glass; Serialized 4130652 Mfr # GLA9PK-AV
COMPARE
Amber 1000 PEAK style
₹154,000.00INR / Each
QLA Footed Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL; Amber Plastic 4130660 Mfr # PLA900-AV
COMPARE
Amber 1000 Footed
₹105,000.00INR / Each
Showing 1- 8 of 8