QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Amber Glass; Serialized
Representative image only.
QLA Standard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Amber Glass; Serialized
| Mfr: GLA900-AV| Item: 4130629
QLA vessels provide a more precise and consistent dissolution experience
₹88,700.00INR / Each
₹88,700.00INR / Each
Product Description
Key Features
  • Compatible with Agilent, VanKel, and Varian dissolution apparatus
  • Standard vessel
  • Amber glass is ideal for light-sensitive compounds
  • 1000 mL capacity
  • A unique serial number is acid etched on the vessel
More About this Item
Vessels are manufactured using a unique molded process which provides a precise and consistent inside diameter and spherical radius resulting in more reproducible dissolution test results.
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.
.
Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorAmber
  • Special FeaturesStandard
  • DescriptionStandard Vessel Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 1000 mL, Amber Glass; Serialized