QLA Sotax-Compatible Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is acid etched on each vessel Glass vessels offer good thermal conductivity Highly resistance to scratching
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom.
76,100.00 - ₹117,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Color Material
Availability Pricing
QLA Standard Vessel for Sotax, 1000 mL, Amber Glass; Serialized 4130627 Mfr # GLA900-AST
COMPARE
1000 Amber Glass
₹117,000.00INR / Each
QLA Standard Vessel for Sotax, 1000 mL, Clear Glass; Serialized 4130639 Mfr # GLA900-ST
COMPARE
1000 Clear Glass
₹76,100.00INR / Each