QLA Standard Vessel for Sotax, 1000 mL, Amber Glass; Serialized

Representative image only.

QLA Standard Vessel for Sotax, 1000 mL, Amber Glass; Serialized

| Mfr: GLA900-AST| Item: 4130627

QLA vessels provide more precise and consistent dissolution results

₹117,000.00INR / Each

₹117,000.00INR / Each
Product Description

Key Features

  • Unique serial number is acid etched on each vessel Glass vessels offer good thermal conductivity Highly resistance to scratching
  • Amber glass is ideal for light-sensitive compounds

More About this Item

Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom.

.

Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • ColorAmber
  • DescriptionStandard Vessel for Sotax, 1000 mL, Amber Glass; Serialized
Reviews
Q&A