QLA MultiDose®-Compatible Shafts
QLA Shafts for dissolution testing with your MultiDose® apparatus
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
These QLA shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with MultiDose® test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. Shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
64,300.00 - ₹82,200.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (mm) Material Description
Availability Pricing
QLA Push-On Style Conical Basket Shaft for MultiDose®, 19"L, 316 SS; Serialized 4130187 Mfr # BSKSHT19-ZM2
COMPARE
483 316 stainless steel Push-On Style Conical Basket Shaft for MultiDose®, 19"L, 316 SS; Serialized
₹82,200.00INR / Each
QLA Push-On Style Conical Basket Shaft for MultiDose®, 15"L, 316 SS; Serialized 4130191 Mfr # BSKSHT-ZM2
COMPARE
381 316 stainless steel Push-On Style Conical Basket Shaft for MultiDose®, 15"L, 316 SS; Serialized
₹64,300.00INR / Each