QLA MultiDose®-Compatible Baskets
QLA Baskets for dissolution testing with your MultiDose® apparatus
  • Made of 316 electropolished stainless steel All baskets and shafts are laser marked with unique serial numbers
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with MultiDose® test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
38,600.00 - ₹59,100.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Material Mesh Size Description
Availability Pricing
QLA Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized 4130153 Mfr # BSK040-ZM
COMPARE
316 stainless steel 40 Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized
₹38,600.00INR / Each
QLA Precision Length Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized 4130154 Mfr # BSK040-ZM2
COMPARE
316 stainless steel 40 Precision Length Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized
₹59,100.00INR / Each