QLA Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized
Representative image only.
QLA Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized
| Mfr: BSK040-ZM| Item: 4130153
QLA Baskets for dissolution testing with your MultiDose® apparatus
₹38,600.00INR / Each
₹38,600.00INR / Each
Buy It Together
Current Item
QLA Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized
QLA Basket for MultiDose®, 40 Mesh USP, 316 SS; Serialized
4130153
₹38,600.00INR / Each
+
QLA Basket Holder, Universal Compatibility; 6 Position
4130170
₹39,100.00INR / Each
=
Total Price 77700.00 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Made of 316 electropolished stainless steel All baskets and shafts are laser marked with unique serial numbers
  • Economical basket for MultiDose® apparatus
More About this Item
These QLA baskets are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with MultiDose® test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All baskets and shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
.
Specifications
  • Material316 stainless steel
  • Mesh Size40
  • DescriptionBasket for MultiDose®, 40 Mesh USP, 316 SS; Serialized