QLA Hanson SR8-Plus Compatible Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • Achieve reproducible results with precision molded vessels
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.
49,400.00 - ₹95,100.00INR / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Bottle Color Capacity (mL) Special Features
Availability Pricing
QLA Standard Vessel for Hanson SR8-Plus, 2000 mL, Clear Glass; Serialized 4130604 Mfr # GLA02K-HR
COMPARE
Clear 2000 Standard
₹88,500.00INR / Each
QLA Standard Vessel for Hanson SR8-Plus, 500 mL; Clear Glass 4130618 Mfr # GLA500-HR
COMPARE
Clear 500 Standard
₹56,100.00INR / Each
QLA Standard Vessel for Hanson SR8-Plus, 1000 mL, Amber Glass; Serialized 4130625 Mfr # GLA900-AH
COMPARE
Amber 1000 Standard
₹73,600.00INR / Each
QLA Standard Vessel for Hanson SR8-Plus, 1000 mL, Clear Glass; Serialized 4130637 Mfr # GLA900-HR
COMPARE
Clear 1000 Standard
₹49,400.00INR / Each
QLA PEAK Vessel with Ring for Hanson SR8-Plus, 1000 mL, Amber Glass; Serialized 4130650 Mfr # GLA9PK-AH
COMPARE
Amber 1000 PEAK-style
₹95,100.00INR / Each
QLA PEAK Vessel with Ring for Hanson SR8-Plus, 1000 mL, Clear Glass; Serialized 4130654 Mfr # GLA9PK-HR
COMPARE
Clear 1000 PEAK-style
₹70,900.00INR / Each
QLA Footed Vessel for Hanson SR8-Plus, 1000 mL; Amber Plastic 4130658 Mfr # PLA900-AH
COMPARE
Amber 1000 Footed
₹54,700.00INR / Each
QLA Footed Vessel for Hanson SR8-Plus, 1000 mL; Clear Plastic 4130663 Mfr # PLA900-HR
COMPARE
Clear 1000 Footed
₹49,600.00INR / Each
Showing 1- 8 of 8