QLA PEAK Vessel with Ring for Hanson SR8-Plus, 1000 mL, Amber Glass; Serialized
Representative image only.
QLA PEAK Vessel with Ring for Hanson SR8-Plus, 1000 mL, Amber Glass; Serialized
| Mfr: GLA9PK-AH| Item: 4130650
QLA vessels provide more precise and consistent dissolution results
₹95,100.00INR / Each
₹95,100.00INR / Each
Product Description
Key Features
  • Achieve reproducible results with precision molded vessels
  • PEAK vessel with ring
  • Amber glass is ideal for light-sensitive compounds
More About this Item
Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom. Select PEAK vessels with a unique cone shape molded into the bottom to improve hydrodynamics, preventing cone-shaped formations of material at the base of the vessel under the paddle. Footed plastic vessels are an economical alternative to glass. They feature feet which allow them to stand upright independently.
Glass vessels offer good thermal conductivity as well as excellent resistance to scratching. A unique serial number is acid etched on the vessel.
.
PEAK vessels do not comply with USP specifications and cannot be substituted in existing applications.
Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • Bottle ColorAmber
  • Special FeaturesPEAK-style
  • DescriptionPEAK Vessel with Ring for Hanson SR8-Plus, 1000 mL, Amber Glass; Serialized