QLA Erweka-Compatible Shafts
QLA Shafts for dissolution testing with your Erweka apparatus
  • Made of 316 electropolished stainless steel All shafts are laser marked with unique serial numbers
These QLA shafts are USP compliant for Apparatus 1 (or Method 1) dissolution testing and compatible with Erweka test apparatus. All are crafted from 316 electropolished stainless steel for strength, smoothness and durability. All shafts are laser marked with unique serial numbers and include Certificates of Conformance that they are USP compliant.
60,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Shaft Length (in) Description
Availability Pricing
QLA Spring Clip Style Basket Shaft for Erweka, 15''L, 10 mm OD, 316 SS; Serialized 4130175 Mfr # BSKSEW-15
COMPARE
15 Spring Clip Style Basket Shaft for Erweka, 15''L, 10 mm OD, 316 SS; Serialized
₹60,300.00INR / Each
QLA Spring Clip Style Basket Shaft for Erweka, 19''L, 10 mm OD, 316 SS; Serialized 4130176 Mfr # BSKSEW-19
COMPARE
19 Spring Clip Style Basket Shaft for Erweka, 19''L, 10 mm OD, 316 SS; Serialized
₹60,300.00INR / Each