QLA Dissolution Filter Discs for Distek Baths
Achieve reproducibility and consistency from test to test with QLA dissolution filters
  • Filter discs are ideal for low-volume sample probes Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds.
11,600.00 - ₹187,000.00INR / Pkg of 1000
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Pore Size (micron) Description
Availability Pricing
QLA Filter Discs, 10 µm, UHMW PE, Distek compatible; 1000/PK 4130902 Mfr # FIL010-DK
COMPARE
10 Filter Discs, 10 µm, UHMW PE, Distek compatible; 1000/PK
₹91,800.00INR / Pkg of 1000
QLA Filter Discs, 10 µm, UHMW PE, Distek compatible; 100/PK 4130903 Mfr # FIL010-DK-a
COMPARE
10 Filter Discs, 10 µm, UHMW PE, Distek compatible; 100/PK
₹11,600.00INR / Pkg of 100
QLA Filter Discs, 20 µm, UHMW PE, Distek compatible; 1000/PK 4130916 Mfr # FIL020-DK
COMPARE
20 Filter Discs, 20 µm, UHMW PE, Distek compatible; 1000/PK
₹114,000.00INR / Pkg of 1000
QLA Filter Discs, 20 µm, UHMW PE, Distek compatible; 100/PK 4130917 Mfr # FIL020-DK-A
COMPARE
20 Filter Discs, 20 µm, UHMW PE, Distek compatible; 100/PK
₹11,600.00INR / Pkg of 100
QLA Filter Discs, 35 µm, UHMW PE, Distek compatible; 1000/PK 4130931 Mfr # FIL035-DK
COMPARE
35 Filter Discs, 35 µm, UHMW PE, Distek compatible; 1000/PK
₹114,000.00INR / Pkg of 1000
QLA Filter Discs, 35 µm, UHMW PE, Distek compatible; 100/PK 4130932 Mfr # FIL035-DK-A
COMPARE
35 Filter Discs, 35 µm, UHMW PE, Distek compatible; 100/PK
₹11,600.00INR / Pkg of 100
QLA Filter Discs, 45 µm, UHMW PE, Distek compatible; 1000/PK 4130949 Mfr # FIL045-DK
COMPARE
45 Filter Discs, 45 µm, UHMW PE, Distek compatible; 1000/PK
₹91,800.00INR / Pkg of 1000
QLA Filter Discs, 45 µm, UHMW PE, Distek compatible; 100/PK 4130950 Mfr # FIL045-DK-A
COMPARE
45 Filter Discs, 45 µm, UHMW PE, Distek compatible; 100/PK
₹11,600.00INR / Pkg of 100
QLA Filter Discs, 70 µm, UHMW PE, Distek compatible; 1000/PK 4130959 Mfr # FIL070-DK
COMPARE
70 Filter Discs, 70 µm, UHMW PE, Distek compatible; 1000/PK
₹187,000.00INR / Pkg of 1000
QLA Filter Discs, 70 µm, UHMW PE, Distek compatible; 100/PK 4130960 Mfr # FIL070-DK-a
COMPARE
70 Filter Discs, 70 µm, UHMW PE, Distek compatible; 100/PK
₹24,000.00INR / Pkg of 100
QLA Filter Discs, 90 µm, UHMW PE, Distek compatible; 1000/PK 4130971 Mfr # FIL090-DK
COMPARE
90 Filter Discs, 90 µm, UHMW PE, Distek compatible; 1000/PK
₹187,000.00INR / Pkg of 1000
QLA Filter Discs, 90 µm, UHMW PE, Distek compatible; 100/PK 4130972 Mfr # FIL090-DK-a
COMPARE
90 Filter Discs, 90 µm, UHMW PE, Distek compatible; 100/PK
₹24,000.00INR / Pkg of 100
Showing 1- 12 of 12