QLA Filter Discs, 70 µm, UHMW PE, Distek compatible; 1000/PK
Representative image only.
QLA Filter Discs, 70 µm, UHMW PE, Distek compatible; 1000/PK
| Mfr: FIL070-DK| Item: 4130959
Achieve reproducibility and consistency from test to test with QLA dissolution filters
₹187,000.00INR / Pkg of 1000
₹187,000.00INR / Pkg of 1000
Product Description
Key Features
  • Filter discs are ideal for low-volume sample probes Keep contaminates out of dissolution samples Guaranteed free of chromium and heavy metal Made of Ultra High Molecular Weight (UHMW) polyethylene
  • 70 µm pore size
  • 1000 per pack
More About this Item
QLA dissolution filters, commonly used in dissolution processes at the ends of the sampling cannula, help keep particulates from backing up into the tubes. Filters are completely inert to most compounds.
.
Specifications
  • MaterialUHMW Polyethylene
  • Pore Size (micron)70
  • DescriptionFilter Discs, 70 µm, UHMW PE, Distek compatible; 1000/PK