QLA Disintegration Testing Beakers, Compatible with Agilent, VanKel, & Varian
High-quality testing with QLA Disintegration Beakers
  • Excellent for use in tablet disintegration systems 900-mL measure etched in glass Manufactured under strict ISO 9001 standards
34,500.00 - ₹38,500.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Material Special Features
Availability Pricing
QLA Disintegration Beaker with Flared Top Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 900 mL; Clear Glass 4130632 Mfr # GLA900-DB
COMPARE
Glass Flared top
₹34,500.00INR / Each
QLA Disintegration Beaker with Flat Top Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 900 mL; Clear Glass 4130642 Mfr # GLA905-DB
COMPARE
Glass Flat top
₹38,500.00INR / Each