QLA Disintegration Beaker with Flat Top Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 900 mL; Clear Glass

Representative image only.

QLA Disintegration Beaker with Flat Top Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 900 mL; Clear Glass

| Mfr: GLA905-DB| Item: 4130642

High-quality testing with QLA Disintegration Beakers

₹38,500.00INR / Each

₹38,500.00INR / Each
Product Description

Key Features

  • Excellent for use in tablet disintegration systems 900-mL measure etched in glass Manufactured under strict ISO 9001 standards
  • Flat top model

More About this Item

.

Not available for sale in the United States.

ISO 9001:2008 Certified Supplier
Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • Max Graduation (mL)900
  • Special FeaturesFlat top
  • DescriptionDisintegration Beaker with Flat Top Compatible with Agilent, VanKel, and Varian Dissolution Apparatus, 900 mL; Clear Glass
Reviews
Q&A