QLA Caleva-Compatible Dissolution Vessels
QLA vessels provide more precise and consistent dissolution results
  • Unique serial number is acid etched on each vessel
Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom.
54,300.00 - ₹80,300.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (mL) Color Material
Availability Pricing
QLA Standard Vessel for Caleva, 1000 mL, Amber Glass; Serialized 4130622 Mfr # GLA900-AC
COMPARE
1000 Amber Glass
₹80,300.00INR / Each
QLA Standard Vessel for Caleva, 1000 mL, Clear Glass; Serialized 4130631 Mfr # GLA900-CA
COMPARE
1000 Clear Glass
₹54,300.00INR / Each