QLA Standard Vessel for Caleva, 1000 mL, Clear Glass; Serialized

Representative image only.

QLA Standard Vessel for Caleva, 1000 mL, Clear Glass; Serialized

| Mfr: GLA900-CA| Item: 4130631

QLA vessels provide more precise and consistent dissolution results

₹54,300.00INR / Each

₹54,300.00INR / Each
Product Description

Key Features

  • Unique serial number is acid etched on each vessel
  • Clear glass

More About this Item

Glass vessels offer good thermal conductivity and are highly resistance to scratching. Standard vessels are manufactured to meet USP physical dimensions and have a perfectly rounded hemispherical bottom.

.

Specifications
  • Capacity (mL)1000
  • MaterialGlass
  • ColorClear
  • DescriptionStandard Vessel for Caleva, 1000 mL, Clear Glass; Serialized
Reviews
Q&A