PYREX® Cyanide Distillation Apparatus
Test for soluble and insoluble cyanides in water
  • Autoclavable
  • Designed for heating by hot plate, flame, oven, microwave or heating mantles
  • In normal service it can be heated many times to 230°C
  • Can be heated one time to 490°C in extreme service
  • Can be cooled to -230°C
Cyanide distillation apparatus tests for soluble and insoluble cyanides in water. Meets EPA and ASTM requirements. Includes round-bottomed flask with inlet tube and funnel opening, adsorber jacket with top, and cold finger with jacket.
356,000.00INR / Case of 1
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Length (cm) Range Resolution
Availability Pricing
Corning 3350-C Cyanide Distillation Apparatus, 500 Ml 3451030 Mfr # 3350-C
COMPARE
₹356,000.00INR / Case of 1