Corning 3350-C Cyanide Distillation Apparatus, 500 Ml
Representative image only.
Corning 3350-C Cyanide Distillation Apparatus, 500 Ml
Corning | Mfr: 3350-C| Item: 3451030
Test for soluble and insoluble cyanides in water
₹356,000.00INR / Case of 1
₹356,000.00INR / Case of 1
Product Description
Key Features
  • Autoclavable
  • Designed for heating by hot plate, flame, oven, microwave or heating mantles
  • In normal service it can be heated many times to 230°C
  • Can be heated one time to 490°C in extreme service
  • Can be cooled to -230°C
More About this Item
Cyanide distillation apparatus tests for soluble and insoluble cyanides in water. Meets EPA and ASTM requirements. Includes round-bottomed flask with inlet tube and funnel opening, adsorber jacket with top, and cold finger with jacket.
Specifications
  • Accessory TypeDistillation Apparatus
  • DescriptionCyanide Distillation Apparatus, 500 Ml