Pyrex Brand 9985 Watch Glasses
These watch glasses provide high chemical durability, preventing contamination of solutions. They are modeled to the same radius of curvature for convenient stacking. The heavy wall an uniform fire-polished edge provide mechanical strength. The outside diameter cannot be controlled within close limits and a slight degree of out-of-round can be expected to occur.
7,900.00 - ₹19,500.00INR / Pkg of 12
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Fits Bottle / Beaker Size (mL) Material Outside Diameter (OD) (cm)
Availability Pricing
Pyrex 9985-65 Brand 9985 watch glass; 65 mm, pack of 12 WW-34507-01 Mfr # 9985-65
COMPARE
150 Glass 6.5
₹7,900.00INR / Pkg of 12
Pyrex 9985-75 Brand 9985 watch glass; 75 mm, pack of 12 WW-34507-02 Mfr # 9985-75
COMPARE
250 Glass 7.5
₹8,400.00INR / Pkg of 12
Pyrex 9985-90 Brand 9985 watch glass; 90 mm, pack of 12 WW-34507-03 Mfr # 9985-90
COMPARE
400 Glass 9
₹9,500.00INR / Pkg of 12
Pyrex 9985-100 Brand 9985 watch glass; 100 mm, pack of 12 WW-34507-04 Mfr # 9985-100
COMPARE
600 Glass 10
₹10,100.00INR / Pkg of 12
Pyrex 9985-125 Brand 9985 watch glass; 125 mm, pack of 12 WW-34507-05 Mfr # 9985-125
COMPARE
1000 Glass 12.5
₹14,300.00INR / Pkg of 12
Pyrex 9985-150 Brand 9985 watch glass; 150 mm, pack of 12 WW-34507-06 Mfr # 9985-150
COMPARE
2000 Glass 15
₹19,500.00INR / Pkg of 12
Pyrex 9985-150 Brand 9985 watch glass; 150 mm, case of 72 WW-34507-26 Mfr # 9985-150
COMPARE
2000 Glass 15
Call For Price
Showing 1- 7 of 7