Pyrex Brand 3160 Dishes
Dishes withstand repeated wet or dry sterilization
  • Easy sorting—tops marked in blue, bottoms marked in white
These flat, clear culture dishes will withstand repeated sterilization(wet or dry). The edges are beaded to provide greater mechanial strength. The bead also provides equal spacing from the walls of the bottom and cover, reducing the capillary action of condensed moisture on the sides. These dishes are not affected chemically or thermally by any of the mehtods commonly employed in laboratories where sterilization is a major factor in routine or in specialized work. The covered dish is not airtight.
The tops are marked in blue enamel and the bottoms in white enamel to make sorting easier. Bottoms also have a triangular, enamel reference point for serial dilutions.
20,000.00 - ₹153,000.00INR / Pkg of 12
12 variations of this product are available.
See All Product Options Below
12 PRODUCT OPTIONS
Compareitem specifications
Outside Diameter (OD) (in)
Availability Pricing
Pyrex 3160-60 Brand 3160 petri dish; 60 x 15 mm, pack of 12 WW-34551-01 Mfr # 3160-60
COMPARE
2 3/8
₹38,000.00INR / Pkg of 12
Pyrex 3160-100 Brand 3160 petri dish; 100 x 10 mm, pack of 12 WW-34551-02 Mfr # 3160-100
COMPARE
3 15/16
₹21,000.00INR / Pkg of 12
Pyrex 3160-101 Brand 3160 petri dish; 100 x 15 mm, pack of 12 WW-34551-03 Mfr # 3160-101
COMPARE
3 15/16
₹21,100.00INR / Pkg of 12
Pyrex 3160-102 Brand 3160 petri dish; 100 x 20 mm, pack of 12 WW-34551-04 Mfr # 3160-102
COMPARE
3 15/16
₹20,000.00INR / Pkg of 12
Pyrex 3160-150 Brand 3160 petri dish; 150 x 15 mm, pack of 12 WW-34551-05 Mfr # 3160-150
COMPARE
5 7/8
₹54,200.00INR / Pkg of 12
Pyrex 3160-152 Brand 3160 petri dish; 150 x 20 mm, pack of 12 WW-34551-06 Mfr # 3160-152
COMPARE
5 7/8
₹50,800.00INR / Pkg of 12
Pyrex 3160-60 Brand 3160 petri dish; 60 x 15 mm, case of 72 WW-34551-21 Mfr # 3160-60
COMPARE
2 3/8
₹153,000.00INR / Case of 72
Pyrex 3160-100 Brand 3160 petri dish; 100 x 10 mm, case of 72 WW-34551-22 Mfr # 3160-100
COMPARE
3 15/16
₹125,000.00INR / Case of 72
Pyrex 3160-101 Brand 3160 petri dish; 100 x 15 mm, case of 72 WW-34551-23 Mfr # 3160-101
COMPARE
3 15/16
₹127,000.00INR / Case of 72
Pyrex 3160-102 Brand 3160 petri dish; 100 x 20 mm, case of 72 WW-34551-24 Mfr # 3160-102
COMPARE
3 15/16
₹120,000.00INR / Case of 72
Pyrex 3160-150 Brand 3160 petri dish; 150 x 15 mm, case of 24 WW-34551-25 Mfr # 3160-150
COMPARE
5 7/8
₹108,000.00INR / Case of 24
Pyrex 3160-152 Brand 3160 petri dish; 150 x 20 mm, case of 24 WW-34551-26 Mfr # 3160-152
COMPARE
5 7/8
₹102,000.00INR / Case of 24
Showing 1- 12 of 12