Pyrex 7078 Disposable Glass Serological Pipettes, Sterile, Bulk-Packed

Disposable glass serological, multi-pack sterile, plugged-end pipettes
  • Features negative graduations
  • Fire-polished tips provide burr-free, even flow rates
  • Reference: ASTM E-714
These sterile serological pipettes are multi-packed in plastic bags for large-volume users. Packs are designed to permit easier access to several pipettes at a time without contaminating the entire case. Size-dependent ISO color-coding is printed directly on the tear strip for easy identification.
Pipettes are calibrated "To Deliver" (TD) with blow out. Long, slender, tapered tips make pipetting faster and easier.
Reference: ASTM E-714
NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.
Call For Price

4 variations of this product are available.

See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Increments (mL) Max Volume (mL)
Availability Pricing
Pyrex 7078-1N Disposable Glass Serological Pipette, 1/0.1 mL, Sterile-Bulk; 1000/Cs WW-34567-23 Mfr # 7078-1N
COMPARE
0.1 1
Call For Price
Pyrex 7078-2N Disposable Glass Serological Pipette, 2 mL, Sterile-Bulk; 700/Cs WW-34567-24 Mfr # 7078-2N
COMPARE
0.01 2
Call For Price
Pyrex 7078-5N Disposable Glass Serological Pipette, 5 mL, Sterile-Bulk; 960/Cs WW-34567-25 Mfr # 7078-5N
COMPARE
0.1 5
Call For Price
Pyrex 7078-10N Disposable Glass Serological Pipette, 10 mL, Sterile-Bulk; 720/Cs WW-34567-26 Mfr # 7078-10N
COMPARE
0.1 10
Call For Price
Showing 1- 4 of 4

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At91-22-61394444
Live chat