Pyrex 7078-2N Disposable Glass Serological Pipette, 2 mL, Sterile-Bulk; 700/Cs

Representative image only.

Pyrex 7078-2N Disposable Glass Serological Pipette, 2 mL, Sterile-Bulk; 700/Cs

Pyrex | Mfr: 7078-2N| Item: 3456724

Disposable glass serological, multi-pack sterile, plugged-end pipettes

Call For Price

Call For Price
Product Description

Key Features

  • Features negative graduations
  • Fire-polished tips provide burr-free, even flow rates
  • Reference: ASTM E-714

More About this Item

These sterile serological pipettes are multi-packed in plastic bags for large-volume users. Packs are designed to permit easier access to several pipettes at a time without contaminating the entire case. Size-dependent ISO color-coding is printed directly on the tear strip for easy identification.
Pipettes are calibrated "To Deliver" (TD) with blow out. Long, slender, tapered tips make pipetting faster and easier.
Reference: ASTM E-714
NOTE: Pipetting by mouth can be hazardous to your health. We suggest using a mechanical pipetting device.

Specifications
  • SterileYes
  • AutoclavableYes
  • Max Volume (µL)2000
  • Min Volume (µL)0
  • Min Volume (mL)0
  • Max Volume (mL)2
  • Max Dispensing (mL)2
  • Graduations (mL)0.01
  • Increments (mL)0.01
  • Overall Length (InchShort)11.41
  • Overall Length (mm)290
  • Body MaterialGlass
  • Color CodeGreen
  • CalibrationTo Deliver (TD)
  • DescriptionDisposableGlassSerologicalPipette,2mL,Sterile-Bulk;700/Cs
Reviews
Q&A