PRO Scientific Micro-Homogenizing Packages
Process microvolumes effectively
  • Narrow generator fits in microtubes—ideal for DNA/RNA extractions
  • Convenient kits provide everything needed to get started
Standard Micro-Homogenizing Package is designed for 500-µL and 1.5-mL tubes with sample sizes up to 0.4 mL. The included small-diameter generator (5 x 75 mm) is best suited for liquid and soft samples.
Deluxe Micro-Homogenizing Package adds a larger saw-tooth generator (7 x 95 mm) to increase versatility. With both generators, 500-µL, 1.5-mL, and 2-mL tubes are compatible. The cutting action of the saw-tooth design can process even tough tissue samples.
Premium Micro-Homogenizing Package builds upon the deluxe kit by adding a stand assembly for hands-free mixing and processing.
487,000.00 - ₹759,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Generator Included Power (VAC)
Availability Pricing
PRO Scientific Standard Micro-Homogenizer Package, 0.5 to 1.5 mL Tubes; 120V 0475113 Mfr # PK-01200S
COMPARE
Small-diameter 120
₹487,000.00INR / Each
PRO Scientific Standard Micro-Homogenizer Package, 0.5 to 1.5 mL Tubes; 220V 0475114 Mfr # PK-02200S
COMPARE
Small-diameter 220
₹512,000.00INR / Each
PRO Scientific Deluxe Micro-Homogenizer Package, 0.5 to 2.0 mL Tubes; 120V 0475116 Mfr # PK-01200D
COMPARE
Saw-tooth and small-diameter 120
Call For Price
PRO Scientific Deluxe Micro-Homogenizer Package, 0.5 to 2.0 mL Tubes; 220V 0475117 Mfr # PK-02200D
COMPARE
Saw-tooth and small-diameter 220
Call For Price
PRO Scientific Premium Micro-Homogenizer Package, 0.5 to 2.0 mL Tubes; 120V 0475118 Mfr # PK-01200P
COMPARE
Saw-tooth and small-diameter 120
₹759,000.00INR / Each
PRO Scientific Premium Micro-Homogenizer Package, 0.5 to 2.0 mL Tubes; 220V 0475119 Mfr # PK-02200P
COMPARE
Saw-tooth and small-diameter 220