Primary Regulators
Provides controlled presssure of gas
A primary regulator is required to adjust the flow rate of fuel gas entering the flame photometer. Choose a regulator from the table according to your fuel type and cylinder valve outlet.
Note: All fuels should be free of heavy hydrocarbon deposits and regulated at the cylinder to 30 psi.
76,700.00 - ₹121,000.00INR / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Sherwood Scientific Propane primary regulator, 5/8" BSP cylinder 0265520 Mfr # 00108234
COMPARE
Propane primary regulator, 5/8" BSP cylinder
₹76,700.00INR / Each
Sherwood Scientific Butane primary regulator, "gaz" type cylinder 0265524 Mfr # 00108732
COMPARE
Butane primary regulator, "gaz" type cylinder
₹81,300.00INR / Each
Sherwood Scientific Primary Regulator, Natural Gas; 7 mm Hosebarb to 1/4" BSP 0265526 Mfr # 41019002
COMPARE
Primary Regulator, Natural Gas; 7 mm Hosebarb to 1/4" BSP
₹121,000.00INR / Each