Sherwood Scientific Propane primary regulator, 5/8" BSP cylinder
Representative image only.
Sherwood Scientific Propane primary regulator, 5/8" BSP cylinder
Sherwood Scientific | Mfr: 00108234| Item: 0265520
Provides controlled presssure of gas
₹76,700.00INR / Each
₹76,700.00INR / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
A primary regulator is required to adjust the flow rate of fuel gas entering the flame photometer. Choose a regulator from the table according to your fuel type and cylinder valve outlet.
Note: All fuels should be free of heavy hydrocarbon deposits and regulated at the cylinder to 30 psi.
1 Year Warranty
Specifications
  • DescriptionPropane primary regulator, 5/8" BSP cylinder