Powerblanket® Ice Insulated Cooling Drum and Pail Blankets
Innovative cooling blanket provides long lasting portable cooling
  • Easily control temperature with ice pack count
  • Advanced insulation directs heat away from vessel keeping temperature sensitive materials cool
  • No power required —portable cooling blankets can be used anywhere
These insulating cooling drum and pail blankets are an easy and cost-effective way to cool large vessels containing temperature sensitive materials. Simply place the ice packs in the pockets on the inside surface of the blanket and place the blanket on to the drum. Modify the number of ice packs used to regulate temperature. Built-in straps securely cinch blanket to the outside of the drum for optimum performance and enhanced portability.
44,300.00 - ₹126,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Width (in) Description
Availability Pricing
Powerblanket PBICE05IP Insulating Cooling Blanket with Ice Packs; 5 Gallon 4202525 Mfr # PBICE05IP
COMPARE
5 gallons 16 Insulating Cooling Blanket with Ice Packs; 5 Gallon
₹44,300.00INR / Each
Powerblanket PBICE55IP Insulating Cooling Blanket with Ice Packs; 55 Gallon 4202528 Mfr # PBICE55IP
COMPARE
55 gallons 36 Insulating Cooling Blanket with Ice Packs; 55 Gallon
₹126,000.00INR / Each