Powerblanket PBICE05IP Insulating Cooling Blanket with Ice Packs; 5 Gallon
Representative image only.
Powerblanket PBICE05IP Insulating Cooling Blanket with Ice Packs; 5 Gallon
Powerblanket | Mfr: PBICE05IP| Item: 4202525
Innovative cooling blanket provides long lasting portable cooling
₹44,300.00INR / Each
₹44,300.00INR / Each
Product Description
Includes
8 ice packs.
Key Features
  • Easily control temperature with ice pack count
  • Advanced insulation directs heat away from vessel keeping temperature sensitive materials cool
  • No power required —portable cooling blankets can be used anywhere
  • Designed to accomidate 5 gallon bucket—blanket is 11.8" by 16"
  • 1 year warranty
More About this Item
These insulating cooling drum and pail blankets are an easy and cost-effective way to cool large vessels containing temperature sensitive materials. Simply place the ice packs in the pockets on the inside surface of the blanket and place the blanket on to the drum. Modify the number of ice packs used to regulate temperature. Built-in straps securely cinch blanket to the outside of the drum for optimum performance and enhanced portability.
1 Year Warranty
Specifications
  • Drum Size5 gallons
  • Width (in)16
  • Length (in)11 51/64
  • DescriptionInsulating Cooling Blanket with Ice Packs; 5 Gallon