Powerblanket® IBC Tote Heaters
Tote heaters maintain optimal heating conditions for sensitive materials
  • Entirely encloses the tote and has a removable top for easy access
  • Durable and weather resistant
  • Features internal thermostats
These tote heaters offer precise temperature control with a programmable digital controller, up to 145°F. Heat is evenly distributed around the tote, reducing hot and cold spots. Rugged vinyl shell is safe to use in temperatures as low as –20°F (–29°F).
517,000.00 - ₹744,000.00INR / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Drum Size Power (VAC)
Availability Pricing
Powerblanket TH275 IBC Tote Heater, 275 Gallons; 120 VAC 4202536 Mfr # TH275
COMPARE
275 gallons 120
₹517,000.00INR / Each
Powerblanket TH275240V IBC Tote Heater, 275 Gallons; 240 VAC 4202537 Mfr # TH275240V
COMPARE
275 gallons 240
₹539,000.00INR / Each
Powerblanket TH330 IBC Tote Heater, 330 Gallons; 120 VAC 4202538 Mfr # TH330
COMPARE
330 gallons 120
₹580,000.00INR / Each
Powerblanket TH330240V IBC Tote Heater, 330 Gallons; 240 VAC 4202539 Mfr # TH330240V
COMPARE
330 gallons 240
Call For Price
Powerblanket TH350 IBC Tote Heater, 350 Gallons; 120 VAC 4202540 Mfr # TH350
COMPARE
350 gallons 120
₹686,000.00INR / Each
Powerblanket TH550 IBC Tote Heater, 550 Gallons; 120 VAC 4202543 Mfr # TH550
COMPARE
550 gallons 120
₹744,000.00INR / Each