Polycarbonate Filter Holders
Designed for the filtration of aqueous solutions
  • Maximum operating pressure is 101 psi
These reusable holders are made of clear, autoclavable polycarbonate with silicone gasket or O-rings. The bell-shaped base protects the filtrate from repeated contamination while following in a receiver.
29,700.00 - ₹79,500.00INR / Pkg of 12
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Body Material Filter Diameter (mm)
Availability Pricing
COMPARE
Polycarbonate 13
₹29,700.00INR / Pkg of 12
COMPARE
Polycarbonate 25
₹29,700.00INR / Pkg of 12
COMPARE
Polycarbonate 50
₹79,500.00INR / Pkg of 5