Pocket Sling Psychrometer
Pocket-sized—take with you anywhere!
To operate psychrometer, withdraw thermometer halfway out of case, then moisten wick. Holding the case as a handle, swing the psychrometer for two minutes. Read wet and dry bulb temperatures, then reinsert thermometer slide into case. The slide and case form a slide rule for calculating RH.
Choose °>F or °>C models. The °>F models are available with either mercury- or spirit-filled thermometers,—use spirit filled model in mercury-free environments. Instructions are included.
12,000.00INR / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max RH Range (%) Min RH Range (%)
Availability Pricing
H-B Instrument B61503-0100 DURAC Compact Sling Psychrometer; 20/120F WW-03322-22 Mfr # 615030100
COMPARE
100 10
₹12,000.00INR / Each
H-B Instrument B61503-0000 DURAC Compact Sling Psychrometer; -5/50C WW-37910-72 Mfr # 615030000
COMPARE
100 10